Product Details

SNP ID
rs200105527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11273445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTCCTCTTCCTCCTCTTCCT[C/T]GCCCGCTGATGACAAGGAGAAGTTA
Phenotype
MIM: 182880 MIM: 182890 MIM: 190232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRM1 PubMed Links

Gene Details

Gene
PRM1
Gene Name
protamine 1
There are no transcripts associated with this gene.

Gene
PRM2
Gene Name
protamine 2
There are no transcripts associated with this gene.

Gene
PRM3
Gene Name
protamine 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021247.2 151 Missense Mutation AAG,GAG K51E NP_067070.2
Gene
TNP2
Gene Name
transition protein 2 (during histone to protamine replacement)
There are no transcripts associated with this gene.

View Full Product Details