Product Details

SNP ID
rs200438465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1911990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTCCACGGGGCCCAGGCCGTGG[C/T]GGAAGGCCAGGGCGCGGAAGCTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HS3ST6 PubMed Links

Gene Details

Gene
HS3ST6
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009606.3 855 Missense Mutation CAC,CGC H210R NP_001009606.3
XM_011522608.2 855 Missense Mutation CAC,CGC H85R XP_011520910.1
XM_011522609.2 855 Missense Mutation CAC,CGC H70R XP_011520911.1

View Full Product Details