Product Details

SNP ID
rs201464467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85903034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAGGATGTGTGACCGGAATGGT[A/G]GTCGGCGGCTTCGACAGTGGCTGAT
Phenotype
MIM: 601565
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRF8 PubMed Links

Gene Details

Gene
IRF8
Gene Name
interferon regulatory factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002163.2 76 Missense Mutation AGT,GGT S7G NP_002154.1
XM_017023199.1 76 UTR 5 XP_016878688.1

View Full Product Details