Product Details

SNP ID
rs200075256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66852798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGACCTACCCGGGCTCTCTGAC[A/G]ACTCCCCCACTCAGTGAGAGTGTCA
Phenotype
MIM: 114770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA7 PubMed Links

Gene Details

Gene
CA7
Gene Name
carbonic anhydrase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014435.1 706 Silent Mutation ACA,ACG T145T NP_001014435.1
NM_005182.2 706 Silent Mutation ACA,ACG T201T NP_005173.1
XM_005256136.3 706 Silent Mutation ACA,ACG T145T XP_005256193.1
XM_011523311.1 706 Silent Mutation ACA,ACG T201T XP_011521613.1
XM_011523312.1 706 Silent Mutation ACA,ACG T145T XP_011521614.1

View Full Product Details