Product Details

SNP ID
rs200241603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69742523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGCCTGGCCTTTTGGGAGAGTC[A/G]CAGCTGAGGGAAGCGCTGATCCTCG
Phenotype
MIM: 613586
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOB1 PubMed Links

Gene Details

Gene
NOB1
Gene Name
NIN1/PSMD8 binding protein 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014062.2 Intron NP_054781.1

View Full Product Details