Product Details

SNP ID
rs200274411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22347590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCACCTCGGCCTCTAGTTCCAGC[A/G]CCCGTGCTCGGTAGGCACCTGTGGC
Phenotype
MIM: 117340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDR2 PubMed Links

Gene Details

Gene
CDR2
Gene Name
cerebellar degeneration related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001802.1 1048 Missense Mutation GCG,GTG A247V NP_001793.1

View Full Product Details