Product Details

SNP ID
rs200570442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:78022688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACCTTACTCCTGGACCAGACCA[C/G]CAGCCACACATCCAGATTAAAAGCC
Phenotype
MIM: 613588
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC3A PubMed Links

Gene Details

Gene
CLEC3A
Gene Name
C-type lectin domain family 3 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244755.1 143 Intron NP_001231684.1
NM_005752.4 143 Missense Mutation ACC,AGC T30S NP_005743.4

View Full Product Details