Product Details
- SNP ID
-
rs200831818
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2223675 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATACGGCAGAAGCCCAGGCCGAAG[A/C]CGGGCGGGAAGCGGGCGAGGGTGCA
- Phenotype
-
MIM: 603022
MIM: 172280
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
E4F1
PubMed Links
Gene Details
- Gene
- E4F1
- Gene Name
- E4F transcription factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001288776.1 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
NP_001275705.1 |
NM_001288778.1 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
NP_001275707.1 |
NM_004424.4 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
NP_004415.3 |
XM_005255155.1 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
XP_005255212.1 |
XM_005255156.1 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
XP_005255213.1 |
XM_006720858.1 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
XP_006720921.1 |
XM_011522402.1 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
XP_011520704.1 |
XM_011522403.1 |
178 |
Missense Mutation |
AGA,AGC |
R34S |
XP_011520705.1 |
XM_017023010.1 |
178 |
Missense Mutation |
GAC,GCC |
D21A |
XP_016878499.1 |
XM_017023011.1 |
178 |
Missense Mutation |
AGA,AGC |
R34S |
XP_016878500.1 |
- Gene
- PGP
- Gene Name
- phosphoglycolate phosphatase
There are no transcripts associated with this gene.
View Full Product Details