Product Details

SNP ID
rs200277145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:82035436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGTCACTGAGAATGAGCACTTT[C/G]TTCTCGGACACAGCATGGATCTGCC
Phenotype
MIM: 109685
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B2 PubMed Links

Gene Details

Gene
HSD17B2
Gene Name
hydroxysteroid 17-beta dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002153.2 184 Missense Mutation TTC,TTG F4L NP_002144.1

View Full Product Details