Product Details

SNP ID
rs200964810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29744779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGGTCTGGGCGGAGAGCACGG[C/T]GGAACAGGCGTTCAGCCAACACAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf54 PubMed Links

Gene Details

Gene
C16orf54
Gene Name
chromosome 16 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175900.3 282 Missense Mutation CAC,CGC H58R NP_787096.2

View Full Product Details