Product Details

SNP ID
rs199757511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19114555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGCCTCTACCATGTCCTGCGG[A/G]GAAGCGGGGGCGCCCGGGCCGAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITPRIPL2 PubMed Links

Gene Details

Gene
ITPRIPL2
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034841.3 624 Missense Mutation AGA,GGA R32G NP_001030013.1

View Full Product Details