Product Details
- SNP ID
-
rs200443167
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89883410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAACGGGGAGAGGTCTGGCTGTGC[G/T]CTCACAGACGCTGTGGCACCAGGGA
- Phenotype
-
MIM: 612326
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TCF25
PubMed Links
Gene Details
- Gene
- TCF25
- Gene Name
- transcription factor 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014972.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
NP_055787.1 |
XM_005256297.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_005256354.1 |
XM_005256298.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_005256355.1 |
XM_005256299.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_005256356.1 |
XM_011522956.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_011521258.1 |
XM_011522957.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_011521259.1 |
XM_011522958.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_011521260.1 |
XM_011522959.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_011521261.1 |
XM_011522960.2 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_011521262.1 |
XM_011522961.2 |
935 |
UTR 5 |
|
|
XP_011521263.1 |
XM_017023047.1 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_016878536.1 |
XM_017023048.1 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_016878537.1 |
XM_017023049.1 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_016878538.1 |
XM_017023050.1 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_016878539.1 |
XM_017023051.1 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_016878540.1 |
XM_017023052.1 |
935 |
Silent Mutation |
GCG,GCT |
A84A |
XP_016878541.1 |
XM_017023053.1 |
935 |
UTR 5 |
|
|
XP_016878542.1 |
XM_017023054.1 |
935 |
UTR 5 |
|
|
XP_016878543.1 |
XM_017023055.1 |
935 |
UTR 5 |
|
|
XP_016878544.1 |
XM_017023056.1 |
935 |
UTR 5 |
|
|
XP_016878545.1 |
XM_017023057.1 |
935 |
UTR 5 |
|
|
XP_016878546.1 |
XM_017023058.1 |
935 |
UTR 5 |
|
|
XP_016878547.1 |
View Full Product Details