Product Details

SNP ID
rs200444255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85678309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTGAGGAAAGTCCCGCTGGTG[G/T]TGATCTCCATCAAGGTCAAGTTATC
Phenotype
MIM: 610609 MIM: 616886
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GINS2 PubMed Links

Gene Details

Gene
GINS2
Gene Name
GINS complex subunit 2
There are no transcripts associated with this gene.

Gene
GSE1
Gene Name
Gse1 coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details