Product Details

SNP ID
rs200900756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:54285102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCAGGCTCGGTGGCCGCGCCG[G/T]CTAAGTTCTCCAAATCGATCTCCTC
Phenotype
MIM: 612985
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IRX3 PubMed Links

Gene Details

Gene
IRX3
Gene Name
iroquois homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024336.2 1501 Missense Mutation GAC,GCC D260A NP_077312.2
XM_005256139.3 1501 Missense Mutation GAC,GCC D260A XP_005256196.1

View Full Product Details