Product Details

SNP ID
rs201095907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74874315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTGTGGCCACCGTGCCCACAG[G/T]TCAGGCAGAAATTGGACGATCCCCG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDR59 PubMed Links

Gene Details

Gene
WDR59
Gene Name
WD repeat domain 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324171.1 2663 Intron NP_001311100.1
NM_001324172.1 2663 Intron NP_001311101.1
NM_030581.3 2663 Missense Mutation AAC,ACC N940T NP_085058.3
XM_005256146.3 2663 Missense Mutation AAC,ACC N959T XP_005256203.1
XM_011523331.2 2663 Missense Mutation AAC,ACC N652T XP_011521633.1
XM_011523332.2 2663 Missense Mutation AAC,ACC N638T XP_011521634.1
XM_017023667.1 2663 Missense Mutation AAC,ACC N688T XP_016879156.1
XM_017023668.1 2663 Missense Mutation AAC,ACC N688T XP_016879157.1
XM_017023669.1 2663 Missense Mutation AAC,ACC N652T XP_016879158.1
XM_017023670.1 2663 Missense Mutation AAC,ACC N638T XP_016879159.1

View Full Product Details