Product Details

SNP ID
rs201322868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15061837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGTCAAAGGAGCTTGGTGTGAT[C/T]CCAATCACGGTATCATTCTGGGGCA
Phenotype
MIM: 615367 MIM: 614244 MIM: 605121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NTAN1 PubMed Links

Gene Details

Gene
NTAN1
Gene Name
N-terminal asparagine amidase
There are no transcripts associated with this gene.

Gene
PDXDC1
Gene Name
pyridoxal dependent decarboxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285444.1 1875 Intron NP_001272373.1
NM_001285445.1 1875 Intron NP_001272374.1
NM_001285447.1 1875 Intron NP_001272376.1
NM_001285448.1 1875 Intron NP_001272377.1
NM_001285449.1 1875 Intron NP_001272378.1
NM_001285450.1 1875 Intron NP_001272379.1
NM_001324019.1 1875 Intron NP_001310948.1
NM_001324020.1 1875 Intron NP_001310949.1
NM_001324021.1 1875 Intron NP_001310950.1
NM_015027.3 1875 Intron NP_055842.2
XM_005255173.1 1875 Intron XP_005255230.1
XM_005255176.2 1875 Intron XP_005255233.1
XM_006720865.2 1875 Intron XP_006720928.2
XM_017023059.1 1875 Intron XP_016878548.1
XM_017023060.1 1875 Intron XP_016878549.1
XM_017023061.1 1875 Intron XP_016878550.1
XM_017023062.1 1875 Intron XP_016878551.1
XM_017023063.1 1875 Intron XP_016878552.1
XM_017023064.1 1875 Intron XP_016878553.1
XM_017023065.1 1875 Intron XP_016878554.1
Gene
RRN3
Gene Name
RRN3 homolog, RNA polymerase I transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301064.1 1875 Silent Mutation GGA,GGG G591G NP_001287993.1
NM_018427.4 1875 Silent Mutation GGA,GGG G621G NP_060897.3
XM_005255376.1 1875 Silent Mutation GGA,GGG G409G XP_005255433.1
XM_005255377.4 1875 Silent Mutation GGA,GGG G409G XP_005255434.1
XM_011522540.2 1875 Silent Mutation GGA,GGG G417G XP_011520842.1
XM_011522541.1 1875 Silent Mutation GGA,GGG G413G XP_011520843.1

View Full Product Details