Product Details

SNP ID
rs199952606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66566650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGGCACAGCGCAACATCTCAG[C/T]GAAGACCGCACCCCGGAAGCACCCC
Phenotype
MIM: 616074 MIM: 607884
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CKLF PubMed Links

Gene Details

Gene
CKLF
Gene Name
chemokine-like factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040138.2 204 Intron NP_001035228.1
NM_016326.3 204 Intron NP_057410.1
NM_016951.3 204 Intron NP_058647.1
NM_181640.2 204 Intron NP_857591.1
NM_181641.2 204 Intron NP_857592.1
Gene
CKLF-CMTM1
Gene Name
CKLF-CMTM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202509.1 204 Intron NP_001189438.1
NM_001204098.1 204 Intron NP_001191027.1
NM_001204099.1 204 Intron NP_001191028.1
Gene
CMTM1
Gene Name
CKLF like MARVEL transmembrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052999.3 204 Missense Mutation GCG,GTG A46V NP_443725.3
NM_181268.2 204 Missense Mutation GCG,GTG A46V NP_851785.2
NM_181269.2 204 Intron NP_851786.1
NM_181270.2 204 Intron NP_851787.1
NM_181271.2 204 Intron NP_851788.1
NM_181272.2 204 Intron NP_851789.1
NM_181283.2 204 Intron NP_851800.1
NM_181296.2 204 Intron NP_851813.1

View Full Product Details