Product Details

SNP ID
rs200411312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56470561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATGAAGTTATTTCGCTCTGAGG[C/T]ACTATTCTTGCTGCTCTCCAACTTC
Phenotype
MIM: 606151 MIM: 615857
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BBS2 PubMed Links

Gene Details

Gene
BBS2
Gene Name
Bardet-Biedl syndrome 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031885.3 2453 Intron NP_114091.3
XM_005256080.2 2453 UTR 3 XP_005256137.1
Gene
OGFOD1
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324357.1 2453 Missense Mutation GCA,GTA A351V NP_001311286.1
NM_001324358.1 2453 Missense Mutation GCA,GTA A297V NP_001311287.1
NM_001324359.1 2453 Missense Mutation GCA,GTA A255V NP_001311288.1
NM_001324360.1 2453 Missense Mutation GCA,GTA A255V NP_001311289.1
NM_001324361.1 2453 Missense Mutation GCA,GTA A297V NP_001311290.1
NM_001324362.1 2453 Missense Mutation GCA,GTA A212V NP_001311291.1
NM_001324363.1 2453 Missense Mutation GCA,GTA A314V NP_001311292.1
NM_018233.3 2453 Missense Mutation GCA,GTA A352V NP_060703.3

View Full Product Details