Product Details
- SNP ID
-
rs200838733
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57741694 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCCTTCCCTGTGTAGAAGAGAT[C/T]GTCGCGCATGCCAGCAACGTGTCCT
- Phenotype
-
MIM: 602703
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KATNB1
PubMed Links
Gene Details
- Gene
- KATNB1
- Gene Name
- katanin regulatory subunit B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005886.2 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
NP_005877.2 |
XM_005255772.4 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_005255829.1 |
XM_006721121.3 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_006721184.1 |
XM_006721122.2 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_006721185.1 |
XM_006721123.3 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_006721186.1 |
XM_011522810.2 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_011521112.1 |
XM_017022860.1 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_016878349.1 |
XM_017022861.1 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_016878350.1 |
XM_017022862.1 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_016878351.1 |
XM_017022863.1 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_016878352.1 |
XM_017022864.1 |
1091 |
Silent Mutation |
ATC,ATT |
I16I |
XP_016878353.1 |
View Full Product Details