Product Details

SNP ID
rs199530196
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85678311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGAGGAAAGTCCCGCTGGTGTT[G/T]ATCTCCATCAAGGTCAAGTTATCCA
Phenotype
MIM: 610609 MIM: 616886
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
GINS2 PubMed Links

Gene Details

Gene
GINS2
Gene Name
GINS complex subunit 2
There are no transcripts associated with this gene.

Gene
GSE1
Gene Name
Gse1 coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details