Product Details

SNP ID
rs199856420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:563879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTGCTATCCCCCGCCTGCCCC[A/C]GGGGAGTTCCCTGAGGCCCACAGCC
Phenotype
MIM: 603267 MIM: 605754
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPN15 PubMed Links

Gene Details

Gene
CAPN15
Gene Name
calpain 15
There are no transcripts associated with this gene.

Gene
LOC107987233
Gene Name
uncharacterized LOC107987233
There are no transcripts associated with this gene.

Gene
NHLRC4
Gene Name
NHL repeat containing 4
There are no transcripts associated with this gene.

Gene
PIGQ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
There are no transcripts associated with this gene.

Gene
PRR35
Gene Name
proline rich 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145270.2 864 Silent Mutation CCA,CCC P195P NP_660313.1
XM_017022959.1 864 Silent Mutation CCA,CCC P195P XP_016878448.1
XM_017022960.1 864 Silent Mutation CCA,CCC P195P XP_016878449.1
XM_017022961.1 864 Intron XP_016878450.1

View Full Product Details