Product Details
- SNP ID
-
rs199598545
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:27226758 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTGACAGATATTGCAGATCTTCA[C/T]CGCGTCGGGGTACGTCTCCCGGATG
- Phenotype
-
MIM: 611917
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KDM8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34940876] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KDM8
- Gene Name
- lysine demethylase 8
There are no transcripts associated with this gene.
- Gene
- NSMCE1
- Gene Name
- NSE1 homolog, SMC5-SMC6 complex component
View Full Product Details