Product Details

SNP ID
rs199892964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:82148800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGTCTCCATTTTCATCTTCTTC[A/C]TAATTGGCATGGTCTCTCTTTCTGG
Phenotype
MIM: 605500
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MPHOSPH6 PubMed Links

Gene Details

Gene
MPHOSPH6
Gene Name
M-phase phosphoprotein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005792.2 577 Silent Mutation NP_005783.2
XM_011522808.2 577 Nonsense Mutation XP_011521110.1
XM_017022852.1 577 Nonsense Mutation XP_016878341.1

View Full Product Details