Product Details
- SNP ID
-
rs200219568
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89708943 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCTTCCGCACAGACACAGATGAT[C/T]CGCAGGGTCCGCACTGCGCCCCAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPATA2L
PubMed Links
Gene Details
- Gene
- SPATA2L
- Gene Name
- spermatogenesis associated 2 like
There are no transcripts associated with this gene.
- Gene
- VPS9D1
- Gene Name
- VPS9 domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004913.2 |
3609 |
Silent Mutation |
CGA,CGG |
R537R |
NP_004904.2 |
XM_005256329.4 |
3609 |
Silent Mutation |
CGA,CGG |
R538R |
XP_005256386.1 |
XM_005256330.4 |
3609 |
Silent Mutation |
CGA,CGG |
R468R |
XP_005256387.1 |
XM_005256331.2 |
3609 |
Silent Mutation |
CGA,CGG |
R372R |
XP_005256388.1 |
XM_006721350.2 |
3609 |
Silent Mutation |
CGA,CGG |
R538R |
XP_006721413.1 |
XM_011523476.2 |
3609 |
Silent Mutation |
CGA,CGG |
R537R |
XP_011521778.1 |
XM_011523477.2 |
3609 |
Silent Mutation |
CGA,CGG |
R504R |
XP_011521779.1 |
XM_011523478.2 |
3609 |
Silent Mutation |
CGA,CGG |
R503R |
XP_011521780.1 |
XM_011523479.2 |
3609 |
Missense Mutation |
GAA,GGA |
E541G |
XP_011521781.1 |
XM_011523480.2 |
3609 |
Silent Mutation |
CGA,CGG |
R468R |
XP_011521782.1 |
- Gene
- VPS9D1-AS1
- Gene Name
- VPS9D1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details