Product Details

SNP ID
rs201513307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11837968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCCTCTTCTTTGATTTTTGGC[C/T]TCTTCTCTGGGCTTTTCCCTGGGGT
Phenotype
MIM: 613746 MIM: 615874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAR4 PubMed Links

Gene Details

Gene
BCAR4
Gene Name
breast cancer anti-estrogen resistance 4 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RSL1D1
Gene Name
ribosomal L1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015659.2 1365 Missense Mutation AAG,AGG K431R NP_056474.2

View Full Product Details