Product Details

SNP ID
rs200221398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71284123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGTTCAGCTGATTCTAGGAGTC[A/G]AACTTCCAACGGTATATGCATTTTG
Phenotype
MIM: 616190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMTR2 PubMed Links

Gene Details

Gene
CMTR2
Gene Name
cap methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099642.1 2348 Nonsense Mutation CGA,TGA R600* NP_001093112.1
NM_001324374.1 2348 Nonsense Mutation CGA,TGA R600* NP_001311303.1
NM_001324377.1 2348 Nonsense Mutation CGA,TGA R600* NP_001311306.1
NM_001324378.1 2348 Nonsense Mutation CGA,TGA R600* NP_001311307.1
NM_001324379.1 2348 Nonsense Mutation CGA,TGA R600* NP_001311308.1
NM_018348.5 2348 Nonsense Mutation CGA,TGA R600* NP_060818.4
XM_011523228.1 2348 Nonsense Mutation CGA,TGA R600* XP_011521530.1
XM_011523229.1 2348 Nonsense Mutation CGA,TGA R600* XP_011521531.1
XM_017023444.1 2348 Nonsense Mutation CGA,TGA R600* XP_016878933.1

View Full Product Details