Product Details

SNP ID
rs201072734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89722702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGCCTGCTTGGTGTGGCTGTCC[A/G]CCAGGACCCCACCTTGTCTCCGTTT
Phenotype
MIM: 608460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS9D1 PubMed Links

Gene Details

Gene
VPS9D1
Gene Name
VPS9 domain containing 1
There are no transcripts associated with this gene.

Gene
VPS9D1-AS1
Gene Name
VPS9D1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF276
Gene Name
zinc finger protein 276
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113525.1 464 Missense Mutation CAC,CGC H126R NP_001106997.1
NM_152287.3 464 Missense Mutation CAC,CGC H51R NP_689500.2
XM_005256324.3 464 Missense Mutation CAC,CGC H126R XP_005256381.1
XM_005256328.3 464 Intron XP_005256385.1
XM_017023889.1 464 Missense Mutation CAC,CGC H126R XP_016879378.1
XM_017023890.1 464 UTR 5 XP_016879379.1

View Full Product Details