Product Details

SNP ID
rs200059061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4813754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTACCTGGGCCAGCTCAGTGA[C/T]GGTGTCAGCGTCCACTGTTGACATG
Phenotype
MIM: 610660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYR1 PubMed Links

Gene Details

Gene
GLYR1
Gene Name
glyoxylate reductase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308096.1 1267 Intron NP_001295025.1
NM_001324096.1 1267 Intron NP_001311025.1
NM_001324097.1 1267 Intron NP_001311026.1
NM_001324098.1 1267 Intron NP_001311027.1
NM_032569.3 1267 Intron NP_115958.2
XM_005255639.4 1267 Intron XP_005255696.2
XM_011522716.2 1267 Missense Mutation ATC,GTC I363V XP_011521018.1
XM_011522717.1 1267 Intron XP_011521019.1
XM_017023786.1 1267 Intron XP_016879275.1
XM_017023787.1 1267 Intron XP_016879276.1
XM_017023788.1 1267 Intron XP_016879277.1
XM_017023789.1 1267 Intron XP_016879278.1

View Full Product Details