Product Details

SNP ID
rs200846717
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57473632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCACCAGCATAGCTGGAGGCTTC[A/G]GCGATGTTCTGGGAACCAGTGATGT
Phenotype
MIM: 608333 MIM: 180663
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DOK4 PubMed Links

Gene Details

Gene
DOK4
Gene Name
docking protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018110.3 1135 Silent Mutation NP_060580.2
XM_005256042.3 1135 Silent Mutation XP_005256099.1
XM_005256043.2 1135 Silent Mutation XP_005256100.1
Gene
POLR2C
Gene Name
RNA polymerase II subunit C
There are no transcripts associated with this gene.

View Full Product Details