Product Details

SNP ID
rs201104896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2461001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGCCCCACCGCATTCCCCAGGC[C/T]AAGCTGGTGGCCATGCTTCAGACAC
Phenotype
MIM: 600227
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf59 PubMed Links

Gene Details

Gene
C16orf59
Gene Name
chromosome 16 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025108.2 317 Nonsense Mutation CAA,TAA Q128* NP_079384.2
XM_011522667.1 317 Nonsense Mutation CAA,TAA Q95* XP_011520969.1
Gene
CCNF
Gene Name
cyclin F
There are no transcripts associated with this gene.

Gene
LOC729652
Gene Name
uncharacterized LOC729652
There are no transcripts associated with this gene.

Gene
MIR6768
Gene Name
microRNA 6768
There are no transcripts associated with this gene.

View Full Product Details