Product Details

SNP ID
rs201267591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29813125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGTCCCAAGGTTCCAGGCGAAG[A/G]GCCTGGCCATTCTGAAGCTGAAACT
Phenotype
MIM: 603213 MIM: 600999 MIM: 605088 MIM: 612033 MIM: 614386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
MAZ
Gene Name
MYC associated zinc finger protein
There are no transcripts associated with this gene.

Gene
MVP
Gene Name
major vault protein
There are no transcripts associated with this gene.

Gene
PAGR1
Gene Name
PAXIP1 associated glutamate rich protein 1
There are no transcripts associated with this gene.

Gene
PRRT2
Gene Name
proline rich transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256442.1 367 Missense Mutation GAG,GGG E24G NP_001243371.1
NM_001256443.1 367 Missense Mutation GAG,GGG E24G NP_001243372.1
NM_145239.2 367 Missense Mutation GAG,GGG E24G NP_660282.2
XM_011545715.2 367 Missense Mutation GAG,GGG E24G XP_011544017.1
XM_011545716.2 367 Missense Mutation GAG,GGG E24G XP_011544018.1
XM_017022887.1 367 Missense Mutation GAG,GGG E24G XP_016878376.1
XM_017022888.1 367 Missense Mutation GAG,GGG E24G XP_016878377.1
XM_017022889.1 367 Missense Mutation GAG,GGG E24G XP_016878378.1

View Full Product Details