Product Details

SNP ID
rs200422452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88639957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGAGCCTGCTGGTGCTGCGCC[A/G]CCGGCCCTGCTCCCGCGACGGCTCG
Phenotype
MIM: 608508 MIM: 604628
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYBA PubMed Links

Gene Details

Gene
CYBA
Gene Name
cytochrome b-245 alpha chain
There are no transcripts associated with this gene.

Gene
IL17C
Gene Name
interleukin 17C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013278.3 528 Missense Mutation CAC,CGC H160R NP_037410.1
Gene
ZC3H18
Gene Name
zinc finger CCCH-type containing 18
There are no transcripts associated with this gene.

View Full Product Details