Product Details

SNP ID
rs200848088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31117796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGAGGGGACCGCCCCATCCCCG[A/G]GCCGCGTCTCTCCGCCGACCCCGGC
Phenotype
MIM: 614901 MIM: 609912
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCKDK PubMed Links

Gene Details

Gene
BCKDK
Gene Name
branched chain ketoacid dehydrogenase kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122957.2 133 Intron NP_001116429.1
NM_001271926.1 133 Intron NP_001258855.1
NM_005881.3 133 Intron NP_005872.2
XM_017022859.1 133 Intron XP_016878348.1
Gene
KAT8
Gene Name
lysine acetyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032188.2 133 Missense Mutation AGC,GGC S39G NP_115564.2
NM_182958.2 133 Missense Mutation AGC,GGC S39G NP_892003.2
XM_011545970.1 133 Intron XP_011544272.1
XM_011545971.1 133 Intron XP_011544273.1

View Full Product Details