Product Details

SNP ID
rs199609511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1093713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCCGCGGGCCGCCGGTTCAGCA[C/T]GATGTGCAGGTAGGTCTCCTTGTAG
Phenotype
MIM: 614147
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF8 PubMed Links

Gene Details

Gene
C1QTNF8
Gene Name
C1q and tumor necrosis factor related protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207419.3 821 Missense Mutation ATG,GTG M183V NP_997302.2
XM_011522483.2 821 Missense Mutation ATG,GTG M183V XP_011520785.1

View Full Product Details