Product Details

SNP ID
rs200094220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30899422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCCCTTTCCCACCAGAAGACCC[A/C]CAGACTGATTCCTCAGTCTCACTTC
Phenotype
MIM: 605847 MIM: 600435
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCL7C PubMed Links

Gene Details

Gene
BCL7C
Gene Name
BCL tumor suppressor 7C
There are no transcripts associated with this gene.

Gene
CTF1
Gene Name
cardiotrophin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142544.1 179 Silent Mutation CCA,CCC P10P NP_001136016.1
NM_001330.3 179 Silent Mutation CCA,CCC P11P NP_001321.1
XM_011545759.2 179 Silent Mutation CCA,CCC P33P XP_011544061.1
XM_011545760.2 179 Silent Mutation CCA,CCC P19P XP_011544062.1
Gene
MIR762
Gene Name
microRNA 762
There are no transcripts associated with this gene.

Gene
MIR762HG
Gene Name
MIR762 host gene
There are no transcripts associated with this gene.

View Full Product Details