Product Details
- SNP ID
-
rs200326283
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2229628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCACTGTGGCCCACATCGTGGTGG[A/G]GGCGGCCTCTCTGGCAGCAGACATC
- Phenotype
-
MIM: 602622
MIM: 603022
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNASE1L2
PubMed Links
Gene Details
- Gene
- DNASE1L2
- Gene Name
- deoxyribonuclease I-like 2
There are no transcripts associated with this gene.
- Gene
- E4F1
- Gene Name
- E4F transcription factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001288776.1 |
564 |
Missense Mutation |
GAG,GGG |
E123G |
NP_001275705.1 |
NM_001288778.1 |
564 |
Missense Mutation |
GAG,GGG |
E123G |
NP_001275707.1 |
NM_004424.4 |
564 |
Missense Mutation |
GAG,GGG |
E123G |
NP_004415.3 |
XM_005255155.1 |
564 |
Missense Mutation |
GAG,GGG |
E176G |
XP_005255212.1 |
XM_005255156.1 |
564 |
Missense Mutation |
GAG,GGG |
E176G |
XP_005255213.1 |
XM_006720858.1 |
564 |
Missense Mutation |
GAG,GGG |
E123G |
XP_006720921.1 |
XM_011522402.1 |
564 |
Missense Mutation |
GAG,GGG |
E176G |
XP_011520704.1 |
XM_011522403.1 |
564 |
Missense Mutation |
GAG,GGG |
E163G |
XP_011520705.1 |
XM_017023010.1 |
564 |
Missense Mutation |
GAG,GGG |
E176G |
XP_016878499.1 |
XM_017023011.1 |
564 |
Missense Mutation |
GAG,GGG |
E163G |
XP_016878500.1 |
View Full Product Details