Product Details

SNP ID
rs201732810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70526668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCCTGCAGCCATGTTTCTGTA[C/T]AACTTAACCTTGCAGAGAGCCACTG
Phenotype
MIM: 606976 MIM: 605592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG4 PubMed Links

Gene Details

Gene
COG4
Gene Name
component of oligomeric golgi complex 4
There are no transcripts associated with this gene.

Gene
SF3B3
Gene Name
splicing factor 3b subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012426.4 223 Silent Mutation TAC,TAT Y4Y NP_036558.3
Gene
SNORD111B
Gene Name
small nucleolar RNA, C/D box 111B
There are no transcripts associated with this gene.

View Full Product Details