Product Details
- SNP ID
-
rs201764849
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2178628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCGTCCAGGATGCTGCCAGTGCT[C/T]TTTTCCGCCGCCGAGTCGCTGCGGG
- Phenotype
-
MIM: 612184
MIM: 606692
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CASKIN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200593605] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASKIN1
- Gene Name
- CASK interacting protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020764.3 |
4250 |
Silent Mutation |
AAA,AAG |
K1406K |
NP_065815.1 |
- Gene
- TRAF7
- Gene Name
- TNF receptor associated factor 7
View Full Product Details