Product Details

SNP ID
rs201830372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:79294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCCTCATAACAACCCACAGGA[C/T]GGTCACCCCCGCTTTGCAGATGAAG
Phenotype
MIM: 156565 MIM: 600928 MIM: 614403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPG PubMed Links

Gene Details

Gene
MPG
Gene Name
N-methylpurine DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015052.2 320 Intron NP_001015052.1
NM_001015054.2 320 Intron NP_001015054.1
NM_002434.3 320 Missense Mutation ACG,ATG T1M NP_002425.2
Gene
NPRL3
Gene Name
NPR3 like, GATOR1 complex subunit
There are no transcripts associated with this gene.

Gene
RHBDF1
Gene Name
rhomboid 5 homolog 1
There are no transcripts associated with this gene.

View Full Product Details