Product Details

SNP ID
rs201642067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1511024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGCTGTTGTGGCGGACCTGCT[C/T]GGGGACGGTGCGTGGCAGTGGGAGA
Phenotype
MIM: 614620 MIM: 611140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT140 PubMed Links

Gene Details

Gene
IFT140
Gene Name
intraflagellar transport 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014714.3 3391 Missense Mutation AAG,GAG K1437E NP_055529.2
XM_005255725.4 3391 Intron XP_005255782.1
XM_005255726.3 3391 Intron XP_005255783.1
XM_006720990.3 3391 Missense Mutation AAG,GAG K1437E XP_006721053.1
XM_006720991.3 3391 Missense Mutation AAG,GAG K1437E XP_006721054.1
XM_006720992.3 3391 Missense Mutation AAG,GAG K648E XP_006721055.1
XM_011522766.2 3391 Missense Mutation AAG,GAG K1355E XP_011521068.1
XM_011522767.2 3391 Missense Mutation AAG,GAG K1112E XP_011521069.1
XM_011522769.2 3391 Intron XP_011521071.1
XM_011522771.2 3391 Intron XP_011521073.1
XM_011522772.2 3391 Intron XP_011521074.1
XM_017023910.1 3391 Missense Mutation AAG,GAG K1437E XP_016879399.1
XM_017023911.1 3391 Missense Mutation AAG,GAG K832E XP_016879400.1
Gene
TELO2
Gene Name
telomere maintenance 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016111.3 3391 Intron NP_057195.2
XM_011522773.2 3391 Intron XP_011521075.1
XM_011522774.2 3391 Intron XP_011521076.1
XM_011522775.2 3391 Intron XP_011521077.1
XM_011522776.2 3391 Intron XP_011521078.1
XM_011522777.2 3391 Intron XP_011521079.1
XM_011522778.2 3391 Intron XP_011521080.1
XM_017023914.1 3391 Intron XP_016879403.1

View Full Product Details