Product Details

SNP ID
rs201766277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58037481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGCTTCTTTGCAGAACTGGCTG[C/T]GGCTTTATGGCTACCTGCCTCAGCC
Phenotype
MIM: 602261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP15 PubMed Links

Gene Details

Gene
MMP15
Gene Name
matrix metallopeptidase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002428.3 957 Missense Mutation CGG,TGG R58W NP_002419.1

View Full Product Details