Product Details

SNP ID
rs202102935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19115186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCCTTCTGCGTGGATGTGCGCG[C/G]GCGGCGTCACCTCTCTGCTACTCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITPRIPL2 PubMed Links

Gene Details

Gene
ITPRIPL2
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034841.3 1255 Missense Mutation GCG,GGG A242G NP_001030013.1

View Full Product Details