Product Details
- SNP ID
-
rs201810566
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57996774 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCGGTGCCGCTGCAGGTCTGACG[C/T]GTACTTGAAGCGTTTCTCGCAGTCT
- Phenotype
-
MIM: 610264
MIM: 613276
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TEPP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3743556] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TEPP
- Gene Name
- testis, prostate and placenta expressed
There are no transcripts associated with this gene.
- Gene
- USB1
- Gene Name
- U6 snRNA biogenesis phosphodiesterase 1
There are no transcripts associated with this gene.
- Gene
- ZNF319
- Gene Name
- zinc finger protein 319
View Full Product Details