Product Details

SNP ID
rs201684326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30927337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCCACACACAGCTGTGTGCCT[C/T]TTGTGTGGGGAGGCTGGGAAGGAGG
Phenotype
MIM: 609085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL19 PubMed Links

Gene Details

Gene
FBXL19
Gene Name
F-box and leucine rich repeat protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099784.2 291 Silent Mutation CTC,CTT L89L NP_001093254.2
NM_001282351.1 291 UTR 5 NP_001269280.1
Gene
FBXL19-AS1
Gene Name
FBXL19 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details