Product Details

SNP ID
rs201748669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:54284246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTCAACCTCGGGGTTTCTTAC[C/T]GCCTGGGCACGGGCTGGAAAGCTGT
Phenotype
MIM: 612985
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRX3 PubMed Links

Gene Details

Gene
IRX3
Gene Name
iroquois homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024336.2 2173 Missense Mutation CAG,CGG Q484R NP_077312.2
XM_005256139.3 2173 Missense Mutation CAC,CGC H484R XP_005256196.1

View Full Product Details