Product Details

SNP ID
rs201778896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30782069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCTGAGATGCTCCAGGAGGAC[A/G]GAAGAGCTCTTCCCCAGCTCTGGAC
Phenotype
MIM: 607700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF40 PubMed Links

Gene Details

Gene
RNF40
Gene Name
ring finger protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207033.1 2467 Intron NP_001193962.1
NM_001207034.1 2467 Intron NP_001193963.1
NM_001286572.2 2467 Intron NP_001273501.1
NM_014771.3 2467 Intron NP_055586.1
XM_011545997.1 2467 Intron XP_011544299.1
Gene
ZNF629
Gene Name
zinc finger protein 629
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080417.1 2467 Silent Mutation TCC,TCT S753S NP_001073886.1
XM_005255222.3 2467 Silent Mutation TCC,TCT S739S XP_005255279.1

View Full Product Details