Product Details

SNP ID
rs201786605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19707113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTTGCAATCGTGCTGGCGGGGC[A/G]GCTGAACGAAGGGGACAGGTTTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf62 PubMed Links

Gene Details

Gene
C16orf62
Gene Name
chromosome 16 open reading frame 62
There are no transcripts associated with this gene.

Gene
IQCK
Gene Name
IQ motif containing K
There are no transcripts associated with this gene.

Gene
KNOP1
Gene Name
lysine rich nucleolar protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012991.2 1433 Intron NP_001013009.2
XM_005255317.4 1433 Intron XP_005255374.1
XM_006721046.3 1433 Missense Mutation CGC,TGC R392C XP_006721109.1
XM_011545855.1 1433 Missense Mutation CGC,TGC R452C XP_011544157.1
XM_017023231.1 1433 Intron XP_016878720.1
XM_017023232.1 1433 Missense Mutation CGC,TGC R369C XP_016878721.1

View Full Product Details