Product Details

SNP ID
rs202046694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2530736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGAACAGCCAGGGAAGAACCAC[C/T]TGCCTGGGCAGGGCCTCGCCTGAGG
Phenotype
MIM: 611113 MIM: 605213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMDHD2 PubMed Links

Gene Details

Gene
AMDHD2
Gene Name
amidohydrolase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145815.1 1859 Silent Mutation CTG,TTG L588L NP_001139287.1
NM_015944.3 1859 Intron NP_057028.2
XM_017023262.1 1859 UTR 3 XP_016878751.1
XM_017023263.1 1859 UTR 3 XP_016878752.1
XM_017023264.1 1859 UTR 3 XP_016878753.1
XM_017023265.1 1859 UTR 3 XP_016878754.1
XM_017023266.1 1859 UTR 3 XP_016878755.1
XM_017023267.1 1859 UTR 3 XP_016878756.1
Gene
CEMP1
Gene Name
cementum protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048212.3 1859 Missense Mutation AAG,AGG K113R NP_001041677.1
Gene
MIR3178
Gene Name
microRNA 3178
There are no transcripts associated with this gene.

Gene
PDPK1
Gene Name
3-phosphoinositide dependent protein kinase 1
There are no transcripts associated with this gene.

View Full Product Details