Product Details

SNP ID
rs199904276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62444843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCCTGCTGCTCCACAGGATGCA[C/G]AAGGCTATCAACAGGCTGAGCAGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
METTL2A PubMed Links

Gene Details

Gene
METTL2A
Gene Name
methyltransferase like 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181725.3 Intron NP_859076.3
XM_017024572.1 Intron XP_016880061.1

View Full Product Details