Product Details

SNP ID
rs200380922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42104835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTGCTCCATCAGCGTCTCCAG[A/C]GCCTCGTTGATCAGCTCCCGCTTCA
Phenotype
MIM: 608588 MIM: 602301
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DHX58 PubMed Links

Gene Details

Gene
DHX58
Gene Name
DEXH-box helicase 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024119.2 1713 Silent Mutation NP_077024.2
XM_017025059.1 1713 Silent Mutation XP_016880548.1
XM_017025060.1 1713 Silent Mutation XP_016880549.1
Gene
KAT2A
Gene Name
lysine acetyltransferase 2A
There are no transcripts associated with this gene.

View Full Product Details